Skip to Content
Merck
All Photos(2)

Key Documents

32544

Supelco

Fenbendazole sulfone

VETRANAL®, analytical standard

Synonym(s):

5-Benzenesulfonyl-1H-benzoimidazol-2-yl)carbamic acid methyl ester, Methyl 5-(phenylsulfonyl)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
Beilstein:
703211
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)(=O)c3ccccc3

InChI

1S/C15H13N3O4S/c1-22-15(19)18-14-16-12-8-7-11(9-13(12)17-14)23(20,21)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

UAFDGCOOJPIAHN-UHFFFAOYSA-N

General description

Chemical structure: benzimidazole

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T G Watson et al.
Research in veterinary science, 38(2), 231-233 (1985-03-01)
The pharmacokinetics of oxfendazole (OFZ) in red deer (Cervus elaphus) was examined. OFZ, administered per os at 4.53 mg kg-1, was extracted in ether from plasma and identified and concentrations estimated by high pressure liquid chromatography. Irrespective of whether the
S E Marriner et al.
American journal of veterinary research, 42(7), 1143-1145 (1981-07-01)
Pharmacokinetics of oxfendazole and its sulfone metabolite were determined in 6 sheep. Oxfendazole achieved mean peak plasma concentrations of 0.76 micrograms/ml at 30 hours after oral administration of oxfendazole (10 mg/kg of body weight), and concentrations were detectable for up
J Landuyt et al.
Biomedical chromatography : BMC, 7(2), 78-81 (1993-03-01)
A high performance liquid chromatographic (HPLC) method for the determination of the anthelminthic pro-benzimidazole febantel and its major metabolites in lamb plasma has been developed. Samples were extracted after addition of albendazole as internal standard, NH4OH and distilled diethyl ether.
P Viviani et al.
Journal of veterinary pharmacology and therapeutics, 41(3), 476-484 (2018-02-22)
Parasitic diseases have a significant impact on livestock production. Nematodicidal drugs, such as fenbendazole (FBZ) or its oxidized metabolite oxfendazole (OFZ), can be used along with the trematodicidal triclabendazole (TCBZ), to broaden the spectrum of anthelmintic activity. However, co-exposure to
Francesca Accioni et al.
Journal of agricultural and food chemistry, 67(1), 520-530 (2018-12-06)
In this work, a restricted access volatile supramolecular solvent (RAM-VOL-SUPRAS) directly synthesized in milk is proposed for the first time for the simultaneous extraction and cleanup of benzimidazole anthelmintic drugs in milk meant for human consumption. The RAM-VOL-SUPRAS was formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service