Skip to Content
Merck
All Photos(2)

Key Documents

26211-U

Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock

1010TLL, PTFE Luer lock (with slots), volume 10 mL, needle size (not included)

Synonym(s):

Precision GASTIGHT Syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500
NACRES:
NB.03

material

PTFE Luer lock (with slots)

needle size

(not included)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 81627

needle L

, n/a

volume

10 mL

fitting

luer lock (without slots)

compatibility

for use with Hamilton 1010TLL (PTFE Luer lock)

Looking for similar products? Visit Product Comparison Guide

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service