Skip to Content
Merck
All Photos(2)

Key Documents

MABF3072

Sigma-Aldrich

Anti-SARS-CoV RBD Antibody, clone m396

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

m396, monoclonal

mol wt

calculated mol wt 139.13 kDa
observed mol wt ~34 kDa

purified by

using Protein A

species reactivity

SARS coronavirus

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
neutralization: suitable
western blot: suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

vaccinia virus ... s> S(1489668)

General description

Spike glycoprotein (UniProt: P59594; also known as S glycoprotein, E2, Peplomer protein) is encoded by the S (also known as 2) gene (Gene ID: 1489668) in SARS-CoV-2 virus. The SARS-CoV-2 is a positive-strand RNA virus that causes severe respiratory syndrome in human. The mature SARS-CoV-2 contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). E and M proteins help in viral assembly and N protein is needed for RNA synthesis. The S protein is a single-pass type I, homotrimeric, membrane glycoprotein that is responsible for virus binding and entry into host cell. It is synthesized with a signal peptide (aa 1-12), which is subsequently cleaved off to generate the mature protein that contains an extracellular domain (aa 13-1213), a transmembrane domain (aa 1214-1234), and a cytoplasmic domain 1235-1273). The S protein is further processed into S1 (aa 13-685) and S2 (aa 686-1273) subunits by host cell furin. The presence of a furin polybasic cleavage site in S protein from SARS-CoV-2 sets it apart from S protein in SARS-CoV that possesses a monobasic S1/S2 cleavage site. The S1 subunit has the receptor binding domain (RBD; aa 319-541) that mediates entry of SARS-CoV-2 into sensitive cells through the peptidase domain of host Angiotensin-converting enzyme 2 (ACE2) with high affinity (KD = 15 nM). The S2 protein, which is reported to be well conserved, is responsible for membrane fusion. Clone m396 is shown to neutralize SARS-CoV-2 virus by disrupting ACE2/RBD binding. It is also effective in neutralizing both SARS-CoV-1 and SARS-CoV-2 pseudoparticles. (Ref.: Ju, B., et al. (2020). Nature. 584(7819); 115-119; Cao, Y., et al. (2020). Cell. 182(1); 73-84).

Specificity

Clone m396 is a human recombinant monoclonal antibody that detects SARS-CoV spike protein. It targets an epitope within the receptor binding domain.

Immunogen

From human IgG phage library panned against the receptor-binding domain (RBD) of the SARS-CoV spike protein.

Application

Quality Control Testing

Evaluated by Western Blotting with His-tagged recombinant receptor binding domain of SARS-CoV spike protein.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant receptor binding domain of SARS-CoV spike protein.

Tested Applications

Neutralizing: A representative lot neutralized SARS-CoV in Neutralizing applications (Zhu, Z., et al. (2007). Proc Natl Acad Sci USA.;104(29):12123-8).

ELISA Analysis: Various dilutions of this antibody detected His-tagged SARS-CoV Receptor Binding Domain (RBD) of SARS-CoV Spike protein.

ELISA Analysis: A representative lot detected SARS-CoV RBD in ELISA applications (Ju, B., et al. (2020). Nature.;584(7819):115-119).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SARS-CoV RBD, clone m396, Cat. No. MABF3072, is a human recombinant monoclonal antibody that detects Spike protein of SARS-CoV and is tested for use in ELISA, Neutralizing, and Western Blotting.

Physical form

Purified human recombinant monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service