Skip to Content
Merck
All Photos(1)

Key Documents

8.41417

Sigma-Aldrich

N-(2-Chloroethyl)-morpholinium chloride

for synthesis

Synonym(s):

N-(2-Chloroethyl)-morpholinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13Cl2NO
CAS Number:
Molecular Weight:
186.08
MDL number:
UNSPSC Code:
12352107
EC Index Number:
222-881-2
NACRES:
NA.22

Quality Level

form

powder

autoignition temp.

365 °C

potency

96 mg/kg LD50, oral (Rat)
1502 mg/kg LD50, skin (Rat)

pH

4.5-5 (20 °C, 10 g/L in H2O)

mp

182-184 °C (decomposition)

storage temp.

2-30°C

InChI

1S/C6H12ClNO.ClH/c7-1-2-8-3-5-9-6-4-8;/h1-6H2;1H

InChI key

NBJHDLKSWUDGJG-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 182 °C
Melting range (upper value): ≤ 186 °C
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service