Skip to Content
Merck
All Photos(1)

Key Documents

8.14123

Sigma-Aldrich

Isophoronediamine

(mixture of stereoisomers) for synthesis

Synonym(s):

Isophoronediamine, 3-Aminomethyl-3,5,5-trimethylcyclohexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2
CAS Number:
Molecular Weight:
170.30
MDL number:
UNSPSC Code:
12352116
EC Index Number:
220-666-8
NACRES:
NA.22

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

380 °C

potency

1030 mg/kg LD50, oral (Rat)

expl. lim.

1.2 % (v/v)

pH

11.6 (20 °C, 10 g/L in H2O)

bp

247 °C/1013 hPa

mp

10 °C

transition temp

flash point 112 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H22N2.C7H8O3S/c1-9(2)4-8(12)5-10(3,6-9)7-11;1-6-2-4-7(5-3-6)11(8,9)10/h8H,4-7,11-12H2,1-3H3;2-5H,1H3,(H,8,9,10)

InChI key

ZBVBXDSQFYHMPD-UHFFFAOYSA-N

Application

  • Flame Retardant Epoxy Compositions: Isophoronediamine was integral in formulating flame-retardant epoxy compositions, demonstrating its utility as a curing agent that significantly reduces flammability. This application is crucial for safety in various industrial sectors including construction and aerospace (Orlov et al., 2022).

Analysis Note

Assay isomers (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.923 - 0.924
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service