Skip to Content
Merck
All Photos(1)

Key Documents

567300

Sigma-Aldrich

SKF-525A, Hydrochloride

Cell permeable. Blocks glibenclamide-sensitive K+ channels. Inhibits neuronal nitric oxide synthase. Also inhibits hepatic drug metabolism by inhibiting the cytochrome P450 system.

Synonym(s):

SKF-525A, Hydrochloride, Proadifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31NO2 · xHCl
CAS Number:
Molecular Weight:
353.50 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C23H31NO2.ClH/c1-4-17-23(20-13-9-7-10-14-20,21-15-11-8-12-16-21)22(25)26-19-18-24(5-2)6-3;/h7-16H,4-6,17-19H2,1-3H3;1H

InChI key

FHIKZROVIDCMJA-UHFFFAOYSA-N

General description

Cell permeable. Blocks glibenclamide-sensitive K+ channels (IC50 = 4.4 µM). Inhibits neuronal nitric oxide synthase (IC50 = 90 µM). Also inhibits hepatic drug metabolism by inhibiting the cytochrome P-450 system. Stimulates endothelial cell prostocyclin production while inhibiting platelet thromboxane synthesis. Potentiates the effect of many drugs in vivo.
Cell permeable. Blocks glibenclamide-sensitive K+ channels. Inhibits neuronal nitric oxide synthase (IC50 = 90 µM). Also inhibits hepatic drug metabolism by inhibiting the cytochrome P450 system. Stimulates endothelial cell prostacyclin production while inhibiting platelet thromboxane synthesis. Potentiates the action of many drugs in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
nNOS
Product does not compete with ATP.
Reversible: no
Target IC50: 90 µM against neuronal nitric oxide synthase

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator(4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Hecker, M., et al. 1994. J. Neurochem. 62, 1524.
Sakuta, H. and Yoneda, I. 1994. Eur. J. Pharmacol.252, 117.
Khan, S., et al. 1993. Biochem. Pharmacol.45, 439.
Boeynaems, J.M., et al. 1986. Prostaglandins32, 145.
Lee, C.H., et al. 1976. J. Pharmacol. Exp. Ther.198, 347.
Grasdalen, J., et al. 1975. FEBS Lett.60, 240.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trista H Wright et al.
Journal of forensic sciences, 58(6), 1558-1562 (2013-08-08)
Two deaths involving 3, 4-methylenedioxypyrovalerone (MDPV) are reported. MDPV is a synthetic cathinone stimulant found in "bath salts" with neurological and cardiovascular toxicity. Biological specimens were analyzed for MDPV by GC/MS and LC/MS. A White man was found dead with
Nathaniel J Pope et al.
Journal of biomedical optics, 25(8), 1-20 (2020-08-14)
Photobiomodulation (PBM) refers to the beneficial effects of low-energy light absorption. Although there is a large body of literature describing downstream physiological benefits of PBM, there is a limited understanding of the molecular mechanisms underlying these effects. At present, the
Tianqi Sheng et al.
Journal of biomedical optics, 25(6), 1-13 (2020-01-09)
Photodynamic therapy (PDT) is a well-established treatment modality for cancer and other malignant diseases; however, quantities such as light fluence and PDT dose do not fully account for all of the dynamic interactions between the key components involved. In particular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service