Skip to Content
Merck
All Photos(1)

Key Documents

535141-M

Millipore

Protease Inhibitor Cocktail V

Animal-Free, lyophilized, for the inhibition of serine and cysteine proteases. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product name

Protease Inhibitor Cocktail Set V, Animal-Free, The Protease Inhibitor Cocktail Set V, Animal-Free controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

storage temp.

−20°C

General description

A cocktail of four protease inhibitors for the inhibition of serine and cysteine proteases, but not metalloproteases. Contains aprotinin that is free of animal products. This cocktail is recommended for use with mammalian cell and tissue extracts. 1X solution will contain the following protease inhibitors:
A cocktail of four protease inhibitors for the inhibition of serine and cysteine proteases, but not metalloproteases. Each vial, when reconstituted with 1 ml H2O, will yield a 100X stock solution. When diluted to 1X the cocktail contains 500 µM AEBSF, HCl (Cat. No. 101500), 150 nM Aprotinin (Cat. No. 616371), 1 µM E-64 (Cat. No. 324890), and 1 µM Leupeptin Hemisulfate (Cat. No. 108975). Available as 1 ml or as 1 set of 10 x 1 ml.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute each vial in 1 ml H₂O to yield a 100X stock solution. Following reconstitution, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service