Skip to Content
Merck
All Photos(1)

Key Documents

5.08161

Sigma-Aldrich

SNC80

Synonym(s):

SNC80, δ Opioid Receptor Agonist, SNC80

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H39N3O2
CAS Number:
Molecular Weight:
449.63
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

0.32 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

cream

solubility

1 M HCl: 100 mM
DMSO: 20 mM

storage temp.

−20°C

InChI

1S/C28H39N3O2/c1-7-17-30-19-22(5)31(20-21(30)4)27(25-11-10-12-26(18-25)33-6)23-13-15-24(16-14-23)28(32)29(8-2)9-3/h7,10-16,18,21-22,27H,1,8-9,17,19-20H2,2-6H3/t21-,22+,27-/m1/s1

InChI key

KQWVAUSXZDRQPZ-UMTXDNHDSA-N

General description

A very potent and selective δ opioid receptor agonist (IC50 = 0.32 µM). Shown to induce dose- and time-dependent antinociception after i.c.v., i.th. and i.p. administration in mice. Widely used in studies of depression and inflammatory injuries. It is also reported to be acted upon heteromeric u-d receptors.

Biochem/physiol Actions

Primary Target
Dela opioid receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pradhan, A., et al., 2013. Mol Pain.9, 8.
Saitoh, A., et al., 2012. Curr. Neuropharmacol.10, 231.
Metcalf. M. et al. 2012. ACS Chem. Neurosci.3, 505.
Bilsky, J., et al. 1995. J. Pharmacol. Exp. Ther.273, 359.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service