Skip to Content
Merck
All Photos(1)

Key Documents

487929-M

Millipore

Nocodazole

InSolution, ≥97%, 50 mM in DMSO, Inhibitor of mammalian cell mitosis via a highly aspecific antimicrotubular activity

Synonym(s):

InSolution Nocodazole, Methyl-(5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl)-carbamate, Methyl-5-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-ylcarbamate, R17934, R17934, Methyl-(5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl)-carbamate, Methyl-5-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3O3S
CAS Number:
Molecular Weight:
301.32
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C14H11N3O3S/c1-20-14(19)17-13-15-9-5-4-8(7-10(9)16-13)12(18)11-3-2-6-21-11/h2-7H,1H3,(H2,15,16,17,19)

InChI key

KYRVNWMVYQXFEU-UHFFFAOYSA-N

General description

Inhibitor of mitosis. Has highly specific antimicrotubular activity for mammalian cells in culture. Promotes tubulin depolymerization. Also acts as a high-affinity ligand for the cancer-related kinases ABL, c-KIT, BRAF, and MEK (Kd<2 M). The solid form of this compound (Cat. No. 487928) is also available.

Biochem/physiol Actions

Primary Target
Microtubules
Secondary Target
Certain kinases

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (10 mg/664 µL) solution of Nocodazole (Cat. No. 487928) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Park, H., et al. 2012. Chem. Med. Chem.7, 53.
Bunz, F., et al. 1998. Science282, 1497.
Fleischman, L.F., et al. 1993. Oncogene8, 771.Pomies, P., and Block, M.R. 1992. Biochem. Biophys. Res. Commun.189, 1429.
Rennison, M.E., et al. 1992. J. Cell Sci.102, 239.
Thyberg, J., and Moskalewski, S. 1992. J. Cell Sci.103, 1167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service