Skip to Content
Merck
All Photos(1)

Key Documents

428016

Millipore

Sodium dodecyl sulfate

High Purity, Designed to reduce electrophoretic anomalies.

Synonym(s):

Sodium n-Dodecyl Sulfate, High Purity, SDS, Sodium Lauryl Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25O4S · Na
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

form

solid

mol wt

micellar wt 18,000

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

62

color

white

CMC

7-10 mM

solubility

water: 100 mg/mL

anion traces

chloride (Cl-): ≤0.05%

cation traces

heavy metals: ≤0.001%
heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Absorbance (3%, H2O, 260 and 280 nm):≤0.1%.
An ionic detergent useful in electrophoretic separation of proteins and lipids (absorbance: ≤0.1% at 280 nm, 3%, H2O; aggregation number: 62). Designed to reduce electrophoretic anomalies. Prepared from SDS (Cat. No. 428015) by a procedure that ensures the removal of contaminating salts and heavy metal ions. Contains C-10 and C-14 isomers. Has a CMC of 7-10 mM and a micellar weight of 18,000.
Designed to reduce electrophoretic anomalies. Prepared from SDS (Cat. No. 428015) by a procedure that ensures the removal of contaminating salts and heavy metal ions. Contains C-10 and C-14 isomers. Aggregation number: 62.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 3 months at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service