Skip to Content
Merck
All Photos(1)

Key Documents

1.15545

Supelco

Knurled nuts for HPLC Columns

for Capillary connections 0.2 mm i.d. 0.5 mm e.d. with PVDF double cones

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115716
NACRES:
SB.52

product name

Knurled nuts for capillary connections 0.2 mm i.d. 0.5 mm e.d. with PVDF double cones,

technique(s)

HPLC: suitable

Quality Level

storage temp.

no temp limit

General description

The package contains 4 knurled nuts.

Analysis Note

visual control: conforms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service