Skip to Content
Merck
All Photos(1)

Key Documents

1.08802

Supelco

Zinc acetate dihydrate

for analysis EMSURE® ACS

Synonym(s):

Zinc acetate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Zn · 2 H2O
CAS Number:
Molecular Weight:
183.48 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
209-170-2
NACRES:
NA.21

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.5-101.0% (complexometric)

form

solid

potency

794 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

6-7 (5% in water)

mp

237 °C

solubility

430 g/L

density

1.74 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.001%
Cd: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.005%
N: ≤0.002%
Na: ≤0.001%
Pb: ≤0.0005%

storage temp.

2-30°C

InChI

1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2

InChI key

BEAZKUGSCHFXIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zinc acetate dihydrate ((CH3COO)2Zn · 2H2O) is hydrated form of zinc acetate. It is bidentate and has covalent character. It is commonly used as a zinc precursor in preparation of zinc oxide nanopowder.

Application

Zinc acetate dihydrate can be used:
  • As a zinc precursor in the synthesis of ZnO nanowires by thermal decomposition method without any catalyst.
  • As a zinc source in the synthesis of ZnO nanoparticles (zinc oxide nanoparticles) with sodium hydroxide by hydrothermal growth method.

Linkage

Replaces: ZX0050-2; ZX0050

Analysis Note

Assay (complexometric): 99.5 - 101.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water): 6.0 - 7.0
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Cd (Cadmium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.001 %
Pb (Lead): ≤ 0.0005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service