Molecular sieves are recommended for achieving the lowest water vapor pressure or absorbing moisture from the atmosphere or gas in a small closed compartment at room temperature.
1.05741
Molecular sieve 0.3 nm
rods ~ 1.6 mm (1/16")
Synonym(s):
Molecular sieve 0.3 nm, Sodium aluminium silicate
About This Item
Recommended Products
Quality Level
form
rod
surface area
800 m2/g
loss
≤10% loss on ignition, 950°C
particle size
~1.6 mm
pore size
0.3 nm
pH
8-11 ( in H2O)
mp
>1600 °C
bulk density
700‑750 kg/m3
storage temp.
no temp limit
SMILES string
[Si](=O)=O.[Al](=O)O[Al]=O
InChI
1S/2Al.O2Si.3O/c;;1-3-2;;;
InChI key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
General description
Application
- in desiccators and drying tubes
- for keeping absolute solvents dry
- for drying gases or solvents in drying columns
- for selective adsorption
Features and Benefits
- Easy to use: practically chemically inert, non-toxic, no disposal problems, dried liquids can be decanted
- High adsorption capacity even with a low water content of the substance to be dried
- High adsorption capacity even at high temperatures
- High adsorption affinity for polar and unsaturated organic molecules (however, H2O is always preferentially adsorbed)
- Selective adsorption: only molecules that can pass through the pores are adsorbed
Analysis Note
Loss on ignition (950 °C): ≤ 10 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 15 %
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Which drying agent should I use if I want to reach as low water vapour pressure as possible in around room temperature? It is a small closed compartment which is initially quite dry so the capacity in not a concern.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service