1.04764
Iodine indicator
Synonym(s):
Indicator for titration of I2 and I-
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
storage temp.
2-30°C
Quality Level
Related Categories
Analysis Note
Solubility in water: passes test
Suitability as indicator (for visual iodine titration): passes test
Suitability as indicator (for visual iodine titration): passes test
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service