Skip to Content
Merck
All Photos(1)

Key Documents

06-333

Sigma-Aldrich

Anti-MAP Kinase 2/Erk2 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

MAP Kinase encoded by the mapk gene

Immunogen

Recombinant mouse ERK2

Application

Anti-MAP Kinase 2/Erk2 Antibody detects level of MAP Kinase 2/Erk2 & has been published & validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
MAP Kinases

Quality

routinely evaluated by immunoblot on mouse 3T3/A31 fibroblasts, human A431 carcinoma cells or rat L6 skeletal fibroblasts

Target description

42kDa

Linkage

Replaces: 04-349

Physical form

0.02M phosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Barber et al.
Journal of immunology (Baltimore, Md. : 1950), 155(3), 1404-1410 (1995-08-01)
LPS-stimulated macrophages (M phi) produce inflammatory mediators that are largely responsible for the pathophysiology associated with septic shock. M phi respond to LPS with rapid protein phosphorylation and dephosphorylation on serine, threonine, and tyrosine residues. If these events are critical
Suppression of c-Myc-induced apoptosis by Ras signalling through PI(3)K and PKB
Kauffmann-Zeh, A., et al
Nature, 385, 544-548 (1997)
Dual phosphorylation and autophosphorylation in mitogen-activated protein (MAP) kinase activation.
Her, J H, et al.
The Biochemical Journal, 296 ( Pt 1), 25-31 (1993)
J D Wright et al.
The Journal of biological chemistry, 270(20), 12085-12093 (1995-05-19)
Although signaling by the epidermal growth factor (EGF) receptor is thought to be dependent on receptor tyrosine kinase activity, it is clear that mitogen-activated protein (MAP) kinase can be activated by receptors lacking kinase activity. Since analysis of the signaling
Granulosa cell expression of G1/S phase cyclins and cyclin-dependent kinases in PMSG-induced follicle growth.
Jennifer D Cannon,Mary Cherian-Shaw,Tara Lovekamp-Swan,Charles L Chaffin
Molecular and cellular endocrinology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service