Skip to Content
Merck
All Photos(2)

Key Documents

05-807

Sigma-Aldrich

Anti-phospho-CREB (Ser133) Antibody, clone 634-2

clone 634-2, Upstate®, from mouse

Synonym(s):

active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, transactivator protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

634-2, monoclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer133)

Gene Information

human ... CREB1(1385)

General description

CREB is a β ZIP transcription factor that activates target genes through cAMP response elements. CREB is able to mediate signals from numerous physiological stimuli, resulting in regulation of a broad array of cellular responses. While CREB is expressed in numerous tissues, it plays a large regulatory role in the nervous system. CREB is believed to play a key role in promoting neuronal survival, precursor proliferation, neurite outgrowth and neuronal differentiation in certain neuronal populations. Additionally, CREB signaling is involved in learning and memory in several organisms. CREB is able to selectively activate numerous downstream genes through interactions with different dimerization partners. CREB is activated by phosphorylation at Ser133 by various signaling pathways including Erk, Ca2+ and stress signaling. Some of the kinases involved in phosphorylating CREB at Ser133 are p90RSK, MSK, CaMKIV and MAPKAPK-2.

Specificity

Broad species cross-reactivity expected, based on sequence homology.
Phospho-CREB (Ser133).

Immunogen

A proprietary immunogen based on the amino acid sequence containing phospho-serine corresponding to residue 133 of human CREB.
Epitope: Ser133

Application

Beadlyte Specificity Assay:
1:500 to 1:32,000 dilutions of this lot detected CREB peptide containing phosphoserine 133. The antibody did NOT recognize the nonphosphorylated peptide of the same sequence

Immunofluorescence / Immunocytochemistry:
This antibody has been reported by an independent laboratory to detect phospho-CREB (Ser133) using immunofluorescence.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-phospho-CREB (Ser133) Antibody, clone 634-2 (mouse monoclonal antibody) validated in ICC, IF, WB to detect phospho-CREB (Ser133) also known as active transcription factor CREB, cAMP responsive element binding protein 1.

Quality

Routinely evaluated by western blot on RIPA lysates from forskolin treated rat PC-12 and mouse 3T3/A31 cells.

Western Blot Analysis:
A 1:1,000 to 1:4,000 dilution of this lot detected phospho-CREB (Ser 133) in RIPA lysates from forskolin treated rat PC-12 and mouse 3T3/A31 cells (Figure A).

Target description

43 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse IgG1κ in buffer containing 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl, 0.07% sodium azide and 30% glycerol. Liquid at -20ºC.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
Human breast carcinoma, forskolin- and FGF-treated SK-N-MC cell extracts.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balapal S Basavarajappa et al.
Hippocampus, 24(7), 808-818 (2014-03-22)
In rodents, many exogenous and endogenous cannabinoids, such as anandamide (AEA) and 2-arachidonyl glycerol (2-AG), have been shown to play an important role in certain hippocampal memory processes. However, the mechanisms by which endogenous AEA regulate this processes are not
A composite element that binds basic helix loop helix and basic leucine zipper transcription factors is important for gonadotropin-releasing hormone regulation of the follicle-stimulating hormone beta gene.
Ciccone, NA; Lacza, CT; Hou, MY; Gregory, SJ; Kam, KY; Xu, S; Kaiser, UB
Molecular Endocrinology null
Marcelo T Marin et al.
The European journal of neuroscience, 30(10), 1931-1940 (2009-11-17)
Learned associations are hypothesized to develop between drug effects and contextual stimuli during repeated drug administration to produce context-specific sensitization that is expressed only in the drug-associated environment and not in a non-drug-paired environment. The neuroadaptations that mediate such context-specific
Rodrigo Molini Leão et al.
Pharmacology, biochemistry, and behavior, 101(3), 434-442 (2012-02-15)
Experimental evidence shows that exposure to stress engenders behavioral sensitization and increases drug-seeking and leads to intense drug taking. However the molecular mechanisms involved in these processes is not well known yet. The present experiments examined the effects of exposure
Yueh-Ting Tsai et al.
Iranian journal of basic medical sciences, 27(6), 706-716 (2024-04-22)
This study assessed the effects of electroacupuncture (EA) stimulation at different frequencies at the Dazhui and Baihui acupoints in the subacute phase after transient global cerebral ischemia (GCI). Rats were subjected to GCI for 25 min, followed by reperfusion for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service