Skip to Content
Merck
All Photos(1)

Documents

D-920

Supelco

N-Desmethyltrimipramine maleate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

N-Desmethyltrimipramine maleate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2 · C4H4O4
CAS Number:
Molecular Weight:
396.48
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

General description

N-Desmethyltrimipramine is a primary blood metabolite of trimipramine. Trimipramine is a tricyclic antidepressant prescribed for treatment of depression, insomnia, and chronic pain or as an adjunctive therapy for alcohol or opioid withdrawal. This Certified Snap-N-Spike® Solution is applicable for use in LC/MS or GC/MS applications for pain prescription monitoring, urine drug testing, clinical toxicology, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Liu et al.
Journal of chromatography. B, Biomedical sciences and applications, 700(1-2), 175-182 (1997-12-09)
A sensitive and stereospecific method for the quantitation of trimipramine enantiomers in human serum was developed. The assay involves the use of a novel mixed-mode disc solid-phase extraction for serum sample clean-up prior to HPLC analysis and is also free
A A Gulaid et al.
Journal of chromatography, 566(1), 228-233 (1991-05-03)
Trimipramine is a tricyclic antidepressant drug often assayed by gas chromatographic or gas chromatographic-mass spectrometry techniques. A high-performance liquid chromatographic method with electrochemical detection is described for the assay of trimipramine and its major metabolites, monodesmethyltrimipramine and 2-hydroxytrimipramine, in plasma.
J Degen et al.
Medizinische Klinik (Munich, Germany : 1983), 88(3), 129-133 (1993-03-15)
The pharmacokinetics of the antidepressants amitriptyline oxide and trimipramine and their major metabolites amitriptyline, nor-triptyline and desmethyltrimipramine, were studied in twelve healthy male subjects (aged from 22 to 62 years) and twelve patients (aged from 25 to 73 years) with
In vitro metabolism of desmethyltrimipramine to yield hydroxylamino compound.
S Ozkirimli et al.
Acta pharmaceutica Suecica, 23(1), 57-59 (1986-01-01)
C B Eap et al.
Therapeutic drug monitoring, 14(5), 380-385 (1992-10-01)
A two-step high-performance liquid chromatography method is described, using a CN column and an alpha 1-acid glycoprotein column, which allows the measurement of the enantiomers of the hydroxy metabolites of trimipramine in plasma of trimipramine-treated patients. Of the four patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service