Skip to Content
Merck
All Photos(3)

Key Documents

840054P

Avanti

Soy PC

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylcholine (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840054P-1g)
pkg of 2 × 100 mg (840054P-200mg)
pkg of 5 × 100 mg (840054P-500mg)
pkg of 1 × 25 mg (840054P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

Phosphatidylcholine (PC) is the main constituent of the plasma membrane. Generally, PC has unsaturated fatty acid, in both from human and vegetable origin. Fatty acid composition of soy PC has 70 % of linolenic acid. Hence, Soy PC has low phase transition temperature in water containing systems.

Application

Soy PC has been used in the preparation of liposomes. Soy PC is suitable for use in the preparation of flexible liposomes (FLUV) and in treating Pheochromocytoma (PC12) cells to analyze its effects on exocytosis kinetics.

Biochem/physiol Actions

Phosphatidylcholine is the main source of phosphocholine, which aids in transforming ceramides into sphingomyelin. Phosphatidylcholine is more prominent in the living tissues. Soy PC is commonly used in cosmetic formulations.

Packaging

5 mL Clear Glass Sealed Ampule (840054P-200mg)
5 mL Clear Glass Sealed Ampule (840054P-25mg)
5 mL Clear Glass Sealed Ampule (840054P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840054P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Cosmetic Science and Technology, 1151(1), 155-155 (2005)
Phospholipid mediated plasticity in exocytosis observed in PC12 cells.
Uchiyama Y, et al.
Brain Research, 1151(1), 46-54 (2007)
Superresolution and fluorescence dynamics evidence reveal that intact liposomes do not cross the human skin barrier.
Dreier J, et al.
PLoS ONE, 11(1), e0146514-e0146514 (2016)
Louiza Bohn Thomsen et al.
Materials (Basel, Switzerland), 12(21) (2019-11-07)
Magnetic nanoparticles have great prospects for drug delivery purposes, as they can be designed with various surface coatings and conjugated with drugs and targeting moieties. They also have a unique potential for precise delivery when guided by magnetic force. The
Targeted antiepidermal growth factor receptor (cetuximab) immunoliposomes enhance cellular uptake in vitro and exhibit increased accumulation in an intracranial model of glioblastoma multiforme.
Mortensen J, et al.
Journal of Drug Delivery, 2013 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service