Skip to Content
Merck
All Photos(2)

Key Documents

830756P

Avanti

17:0 PE

Avanti Research - A Croda Brand

Synonym(s):

PE(17:0/17:0); 110886

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H78NO8P
CAS Number:
Molecular Weight:
720.01
UNSPSC Code:
12352211
NACRES:
NA.25

description

1,2-diheptadecanoyl-sn-glycero-3-phosphoethanolamine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (830756P-200mg)
pkg of 1 × 25 mg (830756P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O

InChI

1S/C39H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-38(41)45-35-37(36-47-49(43,44)46-34-33-40)48-39(42)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36,40H2,1-2H3,(H,43,44)/t37-/m1/s1

InChI key

YSFFAUPDXKTJMR-DIPNUNPCSA-N

Application

17:0 PE has been used as an internal standard for the extraction of lipids.

Packaging

5 mL Amber Glass Screw Cap Vial (830756P-200mg)
5 mL Amber Glass Screw Cap Vial (830756P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of microalgal lipid feedstock by direct-infusion FT-ICR mass spectrometry
Holguin FO and Schaub T
Algal research, 2(1), 43-50 (2013)
Generation and quality control of lipidomics data for the alzheimer?s disease neuroimaging initiative cohort
Barupal DK, et al.
Scientific data, 5, 180263-180263 (2018)
Yasuhiro Horibata et al.
Scientific reports, 10(1), 2845-2845 (2020-02-20)
StarD7 is a phosphatidylcholine (PC)-specific lipid transfer protein essential for the maintenance of mitochondrial PC composition, morphogenesis, and respiration. Here, we studied the role of StarD7 in skeletal myoblast differentiation using mouse myoblast C2C12 cells and human primary myoblasts. Immunofluorescence
Régine Dazzoni et al.
Analytical chemistry, 92(10), 6858-6868 (2020-04-24)
The human nuclear membrane is composed of a double bilayer, the inner membrane being linked to the protein lamina network and the outer nuclear membrane continuous with the endoplasmic reticulum. Nuclear membranes can form large invaginations inside the nucleus; their
Jinfeng Huang et al.
Dose-response : a publication of International Hormesis Society, 18(2), 1559325820914209-1559325820914209 (2020-05-05)
Radiation biodosimeters are required urgently for fast and accurate evaluation of absorbed dose for irradiated individuals. Lipidomics has appeared as a credible technique for identification and quantification of lipid for researching biomarker of diseases. We performed a lipidomic profile on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service