Skip to Content
Merck
All Photos(1)

Key Documents

W326402

Sigma-Aldrich

cis-4-Decenal

≥93%, stabilized, FG

Synonym(s):

(Z)-4-decenal, (Z)-dec-4-enal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH2CHO
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3264
EC Number:
Council of Europe no.:
2297
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.137
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥93%

contains

synthetic alpha-tocopherol as stabilizer

refractive index

n20/D 1.443 (lit.)

bp

78-80 °C/10 mmHg (lit.)

density

0.847 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange; citrus

SMILES string

[H]C(=O)CC\C([H])=C(\[H])CCCCC

InChI

1S/C10H18O/c1-2-3-4-5-6-7-8-9-10-11/h6-7,10H,2-5,8-9H2,1H3/b7-6-

InChI key

CWRKZMLUDFBPAO-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cis-4-decenal is a compound used as a flavoring agent in food products due to its strong, pungent odor (1). The compound enhances the flavor of foods by imparting a savory aroma (2).

Application

cis-4-Decenal is:·
  • one of the volatile compounds present in both Fu-brick tea and Pu-erh tea, which contributes to the aroma of these teas .
  • Found to contribute to the aroma profile of Chinese dry-cured Spanish mackerel .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile composition of Fu-brick tea and Pu-erh tea analyzed by comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry
Shi J, et al.
LWT--Food Science and Technology, 103, 27-33 (2019)
Key aroma compounds of Chinese dry-cured Spanish mackerel (Scomberomorus niphonius) and their potential metabolic mechanisms
Wu S, et al.
Food Chemistry, 342, 128381-128381 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service