Key Documents
BR474400
BRAND® plastic staining trough, Hellendahl pattern
with lid
Synonym(s):
Microscopy, Staining, Staining Accessories
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
material
PMP
feature
with lid
manufacturer/tradename
BRAND 474400
L × W
76 mm × 26 mm , holds 16 slides
General description
The BRAND® Plastic Staining Trough in the Hellendahl pattern is crafted from transparent PMP (polymethylpentene), designed specifically for efficient staining of up to 16 slides, arranged in 8 pairs back-to-back. With dimensions of 76 x 26 mm, this trough provides ample space for optimal staining while ensuring easy access to each slide. Its durable construction allows for reliable use in laboratory settings, making it an essential tool for histology and microscopy applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG