Skip to Content
Merck
All Photos(2)

Key Documents

A41607

Sigma-Aldrich

2-(Trifluoromethyl)aniline

99%

Synonym(s):

α,α,α-Trifluoro-o-toluidine, 2-Aminobenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4NH2
CAS Number:
Molecular Weight:
161.12
Beilstein:
879494
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

170-173 °C (lit.)
68 °C/15 mmHg (lit.)

mp

34 °C (lit.)

density

1.282 g/mL at 25 °C (lit.)

SMILES string

Nc1ccccc1C(F)(F)F

InChI

1S/C7H6F3N/c8-7(9,10)5-3-1-2-4-6(5)11/h1-4H,11H2

InChI key

VBLXCTYLWZJBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 2 Oral

Target Organs

Bone marrow,spleen,Liver

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Torres-Cartas et al.
European journal of medicinal chemistry, 42(11-12), 1396-1402 (2007-05-08)
Mutagenicity is a toxicity endpoint associated with the chronic exposure to chemicals. Aromatic amines have considerable industrial and environmental importance due to their widespread use in industry and their mutagenic capacity. Biopartitioning micellar chromatography (BMC), a mode of micellar liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service