Skip to Content
Merck
All Photos(2)

Key Documents

916013

Sigma-Aldrich

Lithium chloride

greener alternative

anhydrous, 99.95% trace metals basis

Synonym(s):

Lithium monochloride, NSC 327172

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.95% trace metals basis

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

605 °C (lit.)

greener alternative category

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium chloride is used in conjunction with N, N dimethylacetamide (DMA) for synthesis of 5-hydroxymethylfurfural (HMF), a raw material for renewable energy and chemical production, from lignocellulosic biomass. Lithium chloride is also used as a source for synthesis lithium-ion battery materials (e.g. LiFePO4) and to control growth of polyectrolyte multilayers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LiFePO4 Nanoparticles Embedded in a Nanoporous Carbon Matrix: Superior Cathode Material for Electrochemical Energy-Storage Devices..
Wu X L, et al.
Advanced Materials, 21 (25-26), 2710-2714 (2009)
Factors Controlling the Growth of Polyelectrolyte Multilayers.
Dubas S, et al.
Macromolecules, 32(24), 8153-8160 (1999)
Joseph B Binder et al.
Journal of the American Chemical Society, 131(5), 1979-1985 (2009-01-23)
Lignocellulosic biomass is a plentiful and renewable resource for fuels and chemicals. Despite this potential, nearly all renewable fuels and chemicals are now produced from edible resources, such as starch, sugars, and oils; the challenges imposed by notoriously recalcitrant and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service