Skip to Content
Merck
All Photos(1)

Key Documents

777641

Sigma-Aldrich

2,3-Diamino-6-chloropyridine

97%

Synonym(s):

6-Chloropyridine-2,3-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6ClN3
CAS Number:
Molecular Weight:
143.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

131-134 °C

storage temp.

2-8°C

SMILES string

ClC1=NC(N)=C(N)C=C1

InChI

1S/C5H6ClN3/c6-4-2-1-3(7)5(8)9-4/h1-2H,7H2,(H2,8,9)

InChI key

QEIRYIILFUVXAM-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service