Skip to Content
Merck
All Photos(1)

Key Documents

710687

Sigma-Aldrich

Guanosine-13C10 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 13C, ≥95% (CP)

Synonym(s):

GTP-13C10 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C10H14N5Na2O14P3
Molecular Weight:
577.07
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom % 13C

Assay

≥95% (CP)

form

liquid

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+10

storage temp.

−20°C

SMILES string

O=[13C]1[13C](N=[13CH]N2[13C@H]3[13C@H](O)[13C@H](O)[13C@@H]([13CH2]OP(OP(OP(O)(O[Na])=O)(O[Na])=O)(O)=O)O3)=[13C]2N=[13C](N)N1

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and empirical formula are based on the free acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Bartoli et al.
Talanta, 219, 121261-121261 (2020-09-06)
Guanosine tetraphosphate (G4P) and guanosine pentaphosphate (G5P) are signalling nucleotides found in bacteria and photosynthetic eukaryotes that are implicated in a wide-range of processes including stress acclimation, developmental transitions and growth control. Measurements of G4P/G5P levels are essential for studying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service