Skip to Content
Merck
All Photos(2)

Key Documents

678031

Sigma-Aldrich

11-Phosphonoundecanoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23O5P
CAS Number:
Molecular Weight:
266.27
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid

mp

186-190 °C

storage temp.

−20°C

SMILES string

OC(=O)CCCCCCCCCCP(O)(O)=O

InChI

1S/C11H23O5P/c12-11(13)9-7-5-3-1-2-4-6-8-10-17(14,15)16/h1-10H2,(H,12,13)(H2,14,15,16)

InChI key

PPPBZNXJGBLLPM-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pepijn Beekman et al.
Lab on a chip, 19(15), 2526-2536 (2019-07-12)
Tumor-derived extracellular vesicles (tdEVs) are promising blood biomarkers for cancer disease management. However, blood is a highly complex fluid that contains multiple objects in the same size range as tdEVs (30 nm-1 μm), which obscures an unimpeded analysis of tdEVs.
Nicholas J Morris et al.
Nanoscale, 11(37), 17308-17318 (2019-09-13)
Eutectic gallium-indium (EGaIn) is a room temperature liquid metal that can be readily fabricated into nanoparticles which naturally form a thin, passivating gallium oxide shell. These core-shell nanoparticles are of interest for a variety of stimuli-responsive applications, which often utilize

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service