Skip to Content
Merck
All Photos(1)

Key Documents

373885

Sigma-Aldrich

Glycerol propoxylate-block-ethoxylate

average Mn ~5,300

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H22O7
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

<0.3 mmHg ( 20 °C)

description

functionality 2.5

mol wt

average Mn ~5,300

contains

400-800 ppm BHT as inhibitor

composition

ethoxylate, 15%
propoxylate, 85%

density

1.013 g/mL at 25 °C

SMILES string

OCCO.CC(O)CO.OCC(O)CO

InChI

1S/C3H8O3.C3H8O2.C2H6O2/c4-1-3(6)2-5;1-3(5)2-4;3-1-2-4/h3-6H,1-2H2;3-5H,2H2,1H3;3-4H,1-2H2

InChI key

VKVYTKULMKDWLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service