Skip to Content
Merck
All Photos(1)

Key Documents

284416

Sigma-Aldrich

Ethyl hydrocinnamate

99%

Synonym(s):

Ethyl 3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COOC2H5
CAS Number:
Molecular Weight:
178.23
Beilstein:
1910432
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

247-248 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCc1ccccc1

InChI

1S/C11H14O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3

InChI key

JAGZUIGGHGTFHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The transesterifcation reactions of ethyl hydrocinnamate with various alcohols have been studied. It is one of the major components of B. dracunculifolia.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Priya et al.
Indian journal of biochemistry & biophysics, 39(4), 259-263 (2002-08-01)
Use of lipase from Pseudomonas cepacia in transesterifcation reactions of ethyl hydrocinnamate with different alcohols has been examined. Among the alcohols tested, viz., n-butanol, iso-amyl alcohol, benzyl alcohol, n-octanol and 1-phenylethanol, only n-butanol yielded the transesterified product. Among the solvents
Susana Johann et al.
Pharmaceutical biology, 48(4), 388-396 (2010-07-22)
Paracoccidioidomycosis (PCM) is a systemic granulomatous disease caused by Paracoccidioides brasiliensis Almeida (Onygenales) that requires 1-2 years of treatment. In the absence of drug therapy, the disease is usually fatal, highlighting the need for the identification of safer, novel, and
Tanya E Rutan et al.
Journal of agricultural and food chemistry, 66(38), 10053-10066 (2018-09-04)
The effects of two cluster thinning regimes (low and moderate) with Vitis vinifera cv. Pinot noir, in vineyards located in Central Otago, New Zealand, on wine composition, were studied across three consecutive seasons. There were strong correlations between the extent
Eduardo Coelho et al.
Food chemistry, 323, 126826-126826 (2020-04-27)
During beer ageing, endogenous barrel microbes grow spontaneously and transform wort/beer composition, being Dekkera bruxellensis and Saccharomyces cerevisiae among the main contributors to the chemical and sensory profile of aged beer. This work aims at the application of multi-starter cultures
Enbo Xu et al.
Journal of food science, 80(7), C1476-C1489 (2015-06-20)
Enzymatic extrusion, instead of traditional steam cooking, to treat rice is an efficient and alternative pretreatment for Chinese rice wine fermentation. In order to determine the formation of volatiles in enzymatic extrusion-processed rice wine (EE), and to confirm its characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service