Skip to Content
Merck
All Photos(1)

Key Documents

277746

Sigma-Aldrich

Benzoic acid-α-13C

99 atom % 13C

Synonym(s):

13C Labeled benzoic acid, Benzoic acid-(carboxy-13C)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H513CO2H
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

form

solid

bp

249 °C (lit.)

mp

121-125 °C (lit.)

mass shift

M+1

SMILES string

O[13C](=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)/i7+1

InChI key

WPYMKLBDIGXBTP-CDYZYAPPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madalina Neacsu et al.
Molecular nutrition & food research, 61(3) (2016-07-01)
Phytophenols present in cereals are metabolised to compounds that could be partly responsible for the reduced risk of chronic diseases and all-cause mortality associated with fibre-rich diets. The bioavailability, form and in vivo concentrations of these metabolites require to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service