Skip to Content
Merck
All Photos(2)

Key Documents

225754

Sigma-Aldrich

Allylmagnesium bromide solution

1.0 M in diethyl ether

Synonym(s):

(Prop-2-en-1-yl)magnesium bromide, 2-Propenylmagnesium bromide, Allyl magnesium bromide, Allylbromomagnesium, Allylmagnesiumbromide, Bromo-2-propenylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2MgBr
CAS Number:
Molecular Weight:
145.28
Beilstein:
969335
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.851 g/mL at 25 °C

SMILES string

Br[Mg]CC=C

InChI

1S/C3H5.BrH.Mg/c1-3-2;;/h3H,1-2H2;1H;/q;;+1/p-1

InChI key

FEMBXICCJNZMMC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Allylmagnesium bromide solution serves as a selective nucleophile toward aza-aromatic heterocycles.

Application

Allylmagnesium bromide solution can be used in the Grignard reaction, which is a key step to synthesize:
  • 2-(9,10-dihydro-9,10-propanoanthracen-9-yl)-N-methylethanamine (2), a homolog of benzoctamine
  • tarchonanthuslactone
  • (+)-preussin
  • polyhydroxylated quinolizidine alkaloids

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of the antifungal antibiotic (+)-preussin.
Veeresa G, et al.
Tetrahedron, 54(51), 15673-15678 (1998)
An Efficient Asymmetric Synthesis of Tarchonanthuslactone1.
Reddy M, et al.
The Journal of Organic Chemistry, 66(7), 2512-2514 (2001)
Allylmagnesium Bromide as a selective nucleophile toward Aza-aromatic heterocycles
Henry G et al.
Journal of the American Chemical Society, 79, 1245-1249 (1957)
Synthesis of 2-(9, 10-dihydro-9, 10-propanoanthracen-9-yl)-N-methylethanamine via a [4+ 2] cycloaddition.
Karama U, et al.
Molecules (Basel), 15(6), 4201-4206 (2010)
Synthesis of trihydroxy quinolizidine alkaloids: 1, 3-addition reaction of allylmagnesium bromide to a sugar nitrone.
Dhavale DD, et al.
Tetrahedron, 60(13), 3009-3016 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service