GC Analysis of Alkyldiol Enantiomers on Astec® CHIRALDEX™ G-TA
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX G-TA, 40 m x 0.25 mm I.D., 0.12 μm (73034AST)
oven
80 °C (5 min), 5 °C/min to 130 °C
carrier gas
hydrogen, 25 psi
injection
1 μL, 100:1 split
sample
peaks 1-2: propanediol (1.96 and 2.44 min)
peaks 3-4: pentanediol (2.87 and 3.27 min)
peaks 5-6: hexanediol (4.33 and 5.03 min)
peaks 7-8: octanediol (9.68 and 10.18 min)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC