Merck
  • BMI1 regulates androgen receptor in prostate cancer independently of the polycomb repressive complex 1.
BMI1 regulates androgen receptor in prostate cancer independently of the polycomb repressive complex 1.
Nature communications (2018-02-07)
Sen Zhu, Dongyu Zhao, Lin Yan, Weihua Jiang, Jung-Sun Kim, Bingnan Gu, Qipeng Liu, Rui Wang, Bo Xia, Jonathan C Zhao, Gang Song, Wenyi Mi, Rong-Fu Wang, Xiaobing Shi, Hung-Ming Lam, Xuesen Dong, Jindan Yu, Kaifu Chen, Qi Cao
ABSTRACT
BMI1, a polycomb group (PcG) protein, plays a critical role in epigenetic regulation of cell differentiation and proliferation, and cancer stem cell self-renewal. BMI1 is upregulated in multiple types of cancer, including prostate cancer. As a key component of polycomb repressive complex 1 (PRC1), BMI1 exerts its oncogenic functions by enhancing the enzymatic activities of RING1B to ubiquitinate histone H2A at lysine 119 and repress gene transcription. Here, we report a PRC1-independent role of BMI1 that is critical for castration-resistant prostate cancer (CRPC) progression. BMI1 binds the androgen receptor (AR) and prevents MDM2-mediated AR protein degradation, resulting in sustained AR signaling in prostate cancer cells. More importantly, we demonstrate that targeting BMI1 effectively inhibits tumor growth of xenografts that have developed resistance to surgical castration and enzalutamide treatment. These results suggest that blocking BMI1 alone or in combination with anti-AR therapy can be more efficient to suppress prostate tumor growth.
Sigma-Aldrich
MISSION® esiRNA, targeting human RNF2
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MDM2
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
MISSION® esiRNA, targeting human BMI1, COMMD3-BMI1
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)