Merck
  • Tuberin Regulates Prostaglandin Receptor-Mediated Viability, via Rheb, in mTORC1-Hyperactive Cells.
Tuberin Regulates Prostaglandin Receptor-Mediated Viability, via Rheb, in mTORC1-Hyperactive Cells.
Molecular cancer research : MCR (2017-07-16)
Chenggang Li, Xiaolei Liu, Yang Liu, Erik Zhang, Kantha Medepalli, Kouhei Masuda, Na Li, Kathryn A Wikenheiser-Brokamp, Andrew Osterburg, Michael T Borchers, Elizabeth J Kopras, David R Plas, Julia Sun, David N Franz, Jamie K Capal, Maxwell Mays, Yang Sun, David J Kwiatkowski, Anya Alayev, Marina K Holz, Darcy A Krueger, Brian J Siroky, Jane J Yu
ABSTRACT
Tuberous sclerosis complex (TSC) is a tumor-suppressor syndrome affecting multiple organs, including the brain, skin, kidneys, heart, and lungs. TSC is associated with mutations in
Sigma-Aldrich
MISSION® esiRNA, targeting human PTGER3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rheb
Sigma-Aldrich
Sulprostone, ≥95% (HPLC), oil
Sigma-Aldrich
L-798106, ≥98% (HPLC)
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
MISSION® esiRNA, targeting human RHEB