Merck
  • Adherence, fibronectin binding, and induction of cytoskeleton reorganization in cultured human cells by Mycoplasma penetrans.
Adherence, fibronectin binding, and induction of cytoskeleton reorganization in cultured human cells by Mycoplasma penetrans.
Infection and immunity (1996-01-01)
J A Girón, M Lange, J B Baseman
ABSTRACT
Mycoplasma penetrans adhered to cultured human cells, forming clusters that localized to specific areas of the host cell surface. Adherence and cluster formation were inhibited by anti-M. penetrans antibodies, suggesting the involvement of specific adhesin-receptor interactions. Ultrastructural studies showed that after 2 h of infection, mycoplasmas attach to and penetrate the host cell surface. M. penetrans bound selectively to immobilized fibronectin, an interaction which was not inhibited by a 70-kDa fragment containing a heparin-gelatin-binding domain of fibronectin, other matrix glycoproteins, or an RGD tripeptide, suggesting the recognition of other specific binding sites on the fibronectin molecule. A ca. 65-kDa fibronectin-binding protein of M. penetrans was eluted following Sepharose-fibronectin affinity chromatography. Confocal, light, and immunofluorescence microscopy demonstrated that the interaction of M. penetrans with target cells triggers a signal that causes recruitment of several cytoskeletal components, including tubulin and alpha-actinin, and aggregation of phosphorylated proteins. Detergent-soluble mycoplasma proteins with apparent molecular masses of 18, 28, 32, 36, 39, and 41 kDa selectively bound to glutaraldehyde-fixed HEp-2 cells. Our findings offer new insights into understanding the interaction of this human mycoplasma with host target cells.
Sigma-Aldrich
Anti-Tropomyosin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid