Merck
  • Alteration of platelet GPVI signaling in ST-elevation myocardial infarction patients demonstrated by a combination of proteomic, biochemical, and functional approaches.
Alteration of platelet GPVI signaling in ST-elevation myocardial infarction patients demonstrated by a combination of proteomic, biochemical, and functional approaches.
Scientific reports (2016-12-23)
Paula Vélez, Raymundo Ocaranza-Sánchez, Diego López-Otero, Lilian Grigorian-Shamagian, Isaac Rosa, Esteban Guitián, José María García-Acuña, José Ramón González-Juanatey, Ángel García
ABSTRACT
The platelet-specific collagen receptor glycoprotein VI (GPVI) is critical for the formation of arterial thrombosis in vivo. We analyzed GPVI-activated platelets from ST-elevation myocardial infarction (STEMI) patients and matched stable coronary artery disease (SCAD) controls in order to provide novel clues on the degree of involvement of GPVI signaling in the acute event. Firstly, platelets were isolated from systemic venous blood and activated with the GPVI specific agonist CRP (collagen-related peptide). STEMI and SCAD samples were compared by a phosphoproteomics approach. Validations were by immunoblotting in systemic and intracoronary blood from independent cohorts of patients. Twenty-six differentially regulated proteins were identified when comparing CRP-activated systemic platelets from STEMI and SCAD patients, 4 of which were selected for validation studies: PLCɣ2, G6f, SLP-76, and Dok-2. Immunoblot analyses showed these four proteins had higher tyrosine phosphorylation levels in response to CRP in platelets from STEMI patients, being these levels more pronounced at the culprit site of coronary artery occlusion. Moreover, platelet aggregation studies showed a higher response to GPVI agonists in STEMI patients compared to SCAD controls. In conclusion, we show an altered activation state of GPVI signaling in STEMI patients, confirming this receptor as a promising anti-thrombotic target for myocardial infarction.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester, ≥95%, powder
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution