Merck
  • Increased IgA production by B-cells in COPD via lung epithelial interleukin-6 and TACI pathways.
Increased IgA production by B-cells in COPD via lung epithelial interleukin-6 and TACI pathways.
The European respiratory journal (2014-12-30)
Maha Zohra Ladjemi, Marylène Lecocq, Birgit Weynand, Holly Bowen, Hannah J Gould, Jacques Van Snick, Bruno Detry, Charles Pilette
ABSTRACT
Despite their relevance to mucosal defense, production of IgA and the function of lung B-cells remain unknown in chronic obstructive pulmonary disease (COPD). We assessed IgA synthesis in the lungs of COPD (n=28) and control (n=21) patients, and regulation of B-cells co-cultured with in vitro-reconstituted airway epithelium. In COPD lung tissue, synthesis of IgA1 was increased, which led to its accumulation in subepithelial areas. In vitro, the COPD bronchial epithelium imprinted normal human B-cells for increased production of IgA (mainly IgA1) and maturation into CD38(+) plasma cells. These effects were associated with upregulation of TACI (transmembrane activator and CAML interactor) and were observed under resting conditions, while being partly inhibited upon stimulation with cigarette smoke extract. Interleukin (IL)-6 and BAFF (B-cell activating factor)/APRIL (a proliferation-inducing ligand) were upregulated in the COPD epithelium and lung tissue, respectively; the IgA-promoting effect of the COPD bronchial epithelium was inhibited by targeting IL-6 and, to a lower extent, by blocking TACI. These data show that in COPD, the bronchial epithelium imprints B-cells with signals promoting maturation into IgA-producing plasma cells through the action of two epithelial/B-cell axes, namely the IL-6/IL-6 receptor and BAFF-APRIL/TACI pathways, while cigarette smoke partly counteracts this IgA-promoting effect.
Sigma-Aldrich
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody