Merck
  • Splicing function of mitotic regulators links R-loop-mediated DNA damage to tumor cell killing.
Splicing function of mitotic regulators links R-loop-mediated DNA damage to tumor cell killing.
The Journal of cell biology (2015-04-29)
Yihan Wan, Xiaobin Zheng, Haiyang Chen, Yuxuan Guo, Hao Jiang, Xiaonan He, Xueliang Zhu, Yixian Zheng
ABSTRACT
Although studies suggest that perturbing mitotic progression leads to DNA damage and p53 activation, which in turn lead to either cell apoptosis or senescence, it remains unclear how mitotic defects trigger p53 activation. We show that BuGZ and Bub3, which are two mitotic regulators localized in the interphase nucleus, interact with the splicing machinery and are required for pre-mRNA splicing. Similar to inhibition of RNA splicing by pladienolide B, depletion of either BuGZ or Bub3 led to increased formation of RNA-DNA hybrids (R-loops), which led to DNA damage and p53 activation in both human tumor cells and primary cells. Thus, R-loop-mediated DNA damage and p53 activation offer a mechanistic explanation for apoptosis of cancer cells and senescence of primary cells upon disruption of the dual-function mitotic regulators. This demonstrates the importance of understanding the full range of functions of mitotic regulators to develop antitumor drugs.
Sigma-Aldrich
Anti-ZNF207 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.