Merck
  • Imaging Abeta plaques in living transgenic mice with multiphoton microscopy and methoxy-X04, a systemically administered Congo red derivative.
Imaging Abeta plaques in living transgenic mice with multiphoton microscopy and methoxy-X04, a systemically administered Congo red derivative.
Journal of neuropathology and experimental neurology (2002-09-17)
William E Klunk, Brian J Bacskai, Chester A Mathis, Stephen T Kajdasz, Megan E McLellan, Matthew P Frosch, Manik L Debnath, Daniel P Holt, Yanming Wang, Bradley T Hyman
ABSTRACT
The identification of amyloid deposits in living Alzheimer disease (AD) patients is important for both early diagnosis and for monitoring the efficacy of newly developed anti-amyloid therapies. Methoxy-X04 is a derivative of Congo red and Chrysamine-G that contains no acid groups and is therefore smaller and much more lipophilic than Congo red or Chrysamine-G. Methoxy-X04 retains in vitro binding affinity for amyloid beta (Abeta) fibrils (Ki = 26.8 nM) very similar to that of Chrysamine-G (Ki = 25.3 nM). Methoxy-X04 is fluorescent and stains plaques, tangles, and cerebrovascular amyloid in postmortem sections of AD brain with good specificity. Using multiphoton microscopy to obtain high-resolution (1 microm) fluorescent images from the brains of living PSI/APP mice, individual plaques could be distinguished within 30 to 60 min after a single i.v. injection of 5 to 10 mg/kg methoxy-X04. A single i.p. injection of 10 mg/kg methoxy-X04 also produced high contrast images of plaques and cerebrovascular amyloid in PSI/APP mouse brain. Complementary quantitative studies using tracer doses of carbon- 11-labeled methoxy-X04 show that it enters rat brain in amounts that suggest it is a viable candidate as a positron emission tomography (PET) amyloid-imaging agent for in vivo human studies.
Sigma-Aldrich
Methoxy-X04, ≥98% (HPLC)
Sigma-Aldrich
Chrysamine G, ≥95% (HPLC), solid