Merck
  • The relationship between epicardial fat tissue thickness and visceral adipose tissue in lean patients with polycystic ovary syndrome.
The relationship between epicardial fat tissue thickness and visceral adipose tissue in lean patients with polycystic ovary syndrome.
Journal of ovarian research (2015-11-08)
Dilek Arpaci, Aysel Gurkan Tocoglu, Sabiye Yilmaz, Hasan Ergenc, Ali Tamer, Nurgul Keser, Huseyin Gunduz
ABSTRACT
Polycystic ovary syndrome (PCOS) is related to metabolic syndrome, insulin resistance, and cardiovascular metabolic syndromes. This is particularly true for individuals with central and abdominal obesity because visceral abdominal adipose tissue (VAAT) and epicardial adipose tissue (EAT) produce a large number of proinflammatory and proatherogenic cytokines. The present study aimed to determine whether there are changes in VAAT and EAT levels which were considered as indirect predictors for subclinical atherosclerosis in lean patients with PCOS. The clinical and demographic characteristics of 35 patients with PCOS and 38 healthy control subjects were recorded for the present study. Additionally, the serum levels of various biochemical parameters were measured and EAT levels were assessed using 2D-transthoracic echocardiography. There were no significant differences in mean age (p = 0.056) or mean body mass index (BMI) (p = 0.446) between the patient and control groups. However, the body fat percentage, waist-to-hip ratio, amount of abdominal subcutaneous adipose tissue, and VAAT thickness were higher in the PCOS patient group than in the control group. The amounts of EAT in the patient and control groups were similar (p = 0.384). EAT was correlated with BMI, fat mass, waist circumference, and hip circumference but not with any biochemical metabolic parameters including the homeostasis model assessment of insulin resistance index or the levels of triglycerides, low-density lipoprotein cholesterol, and high-density lipoprotein (HDL) cholesterol. However, there was a small positive correlation between the amounts of VAAT and EAT. VAAT was directly correlated with body fat parameters such as BMI, fat mass, and abdominal subcutaneous adipose thickness and inversely correlated with the HDL cholesterol level. The present study found that increased abdominal adipose tissue in patients with PCOS was associated with atherosclerosis. Additionally, EAT may aid in the determination of the risk of atherosclerosis in patients with PCOS because it is easily measured.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
2,3-Dimethylaniline, 99%