Merck
  • Honokiol inhibits the growth of head and neck squamous cell carcinoma by targeting epidermal growth factor receptor.
Honokiol inhibits the growth of head and neck squamous cell carcinoma by targeting epidermal growth factor receptor.
Oncotarget (2015-05-29)
Tripti Singh, Nirzari A Gupta, Su Xu, Ram Prasad, Sadanandan E Velu, Santosh K Katiyar
ABSTRACT
Here, we report the chemotherapeutic effect of honokiol, a phytochemical from Magnolia plant, on human head and neck squamous cell carcinoma (HNSCC). Treatment of HNSCC cell lines from different sub-sites, SCC-1 (oral cavity), SCC-5 (larynx), OSC-19 (tongue) and FaDu (pharynx) with honokiol inhibited their cell viability, which was associated with the: (i) induction of apoptosis, (ii) correction of dysregulatory cell cycle proteins of G0/G1 phase. Honokiol decreased the expression levels of epidermal growth factor receptor (EGFR), mTOR and their downstream signaling molecules. Treatment of FaDu and SCC-1 cell lines with rapamycin, an inhibitor of mTOR pathway, also reduced cell viability of HNSCC cells. Administration of honokiol by oral gavage (100 mg/kg body weight) significantly (P < 0.01-0.001) inhibited the growth of SCC-1 and FaDu xenografts in athymic nude mice, which was associated with: (i) inhibition of tumor cell proliferation, (ii) induction of apoptosis, (iii) reduced expressions of cyclins and Cdks, and (iv) inhibition of EGFR signaling pathway. Molecular docking analysis of honokiol in EGFR binding site indicated that the chemotherapeutic effect of honokiol against HNSCC is mediated through its firm binding with EGFR, which is better than that of gefitinib, a commonly used drug for HNSCC treatment.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human EGFR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Egfr