Merck
  • Delivery of aerosolized liposomal amikacin as a novel approach for the treatment of nontuberculous mycobacteria in an experimental model of pulmonary infection.
Delivery of aerosolized liposomal amikacin as a novel approach for the treatment of nontuberculous mycobacteria in an experimental model of pulmonary infection.
PloS one (2014-09-30)
Sasha J Rose, Mary E Neville, Renu Gupta, Luiz E Bermudez
ABSTRACT
Pulmonary infections caused by nontuberculous mycobacteria (NTM) are an increasing problem in individuals with chronic lung conditions and current therapies are lacking. We investigated the activity of liposomal amikacin for inhalation (LAI) against NTM in vitro as well as in a murine model of respiratory infection. Macrophage monolayers were infected with three strains of Mycobacterium avium, two strains of Mycobacterium abscessus, and exposed to LAI or free amikacin for 4 days before enumerating bacterial survival. Respiratory infection was established in mice by intranasal inoculation with M. avium and allowing three weeks for the infection to progress. Three different regimens of inhaled LAI were compared to inhaled saline and parenterally administered free amikacin over a 28 day period. Bacteria recovered from the mice were analyzed for acquired resistance to amikacin. In vitro, liposomal amikacin for inhalation was more effective than free amikacin in eliminating both intracellular M. avium and M. abscessus. In vivo, inhaled LAI demonstrated similar effectiveness to a ∼25% higher total dose of parenterally administered amikacin at reducing M. avium in the lungs when compared to inhaled saline. Additionally, there was no acquired resistance to amikacin observed after the treatment regimen. The data suggest that LAI has the potential to be an effective therapy against NTM respiratory infections in humans.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Amikacin disulfate salt, potency: 674-786 μg per mg (as amikacin base)
Amikacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
USP
Amikacin sulfate, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Amikacin sulfate, European Pharmacopoeia (EP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material