Merck
  • Elevated serum squalene and cholesterol synthesis markers in pregnant obese women with gestational diabetes mellitus.
Elevated serum squalene and cholesterol synthesis markers in pregnant obese women with gestational diabetes mellitus.
Journal of lipid research (2014-10-11)
Helena E Miettinen, Kristiina Rönö, Saila Koivusalo, Beata Stach-Lempinen, Maritta Pöyhönen-Alho, Johan G Eriksson, Timo P Hiltunen, Helena Gylling
ABSTRACT
We examined serum cholesterol synthesis and absorption markers and their association with neonatal birth weight in obese pregnancies affected by gestational diabetes mellitus (GDM). Pregnant women at risk for GDM (BMI >30 kg/m²) were enrolled from maternity clinics in Finland. GDM was determined from the results of an oral glucose tolerance test. Serum samples were collected at six time-points, one in each trimester of pregnancy, and at 6 weeks, 6 months, and 12 months postpartum. Analysis of serum squalene and noncholesterol sterols by gas-liquid chromatography revealed that in subjects with GDM (n = 22), the serum Δ8-cholestenol concentration and lathosterol/sitosterol ratio were higher (P < 0.05) than in the controls (n = 30) in the first trimester, reflecting increased cholesterol synthesis. Also, subjects with GDM had an increased ratio of squalene to cholesterol (100 × μmol/mmol of cholesterol) in the second (11.5 ± 0.5 vs. 9.1 ± 0.5, P < 0.01) and third (12.1 ± 0.8 vs. 10.0 ± 0.7, P < 0.05) trimester. In GDM, the second trimester maternal serum squalene concentration correlated with neonatal birth weight (r = 0.70, P < 0.001). In conclusion, in obesity, GDM associated with elevated serum markers of cholesterol synthesis. Correlation of maternal serum squalene with neonatal birth weight suggests a potential contribution of maternal cholesterol synthesis to newborn weight in GDM.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Squalene, analytical standard
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
Squalene, ≥98%, liquid
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, ≥70%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Beta-sitosterol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material