Merck
  • Fibrosis progression in nonalcoholic fatty liver vs nonalcoholic steatohepatitis: a systematic review and meta-analysis of paired-biopsy studies.
Fibrosis progression in nonalcoholic fatty liver vs nonalcoholic steatohepatitis: a systematic review and meta-analysis of paired-biopsy studies.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association (2014-04-29)
Siddharth Singh, Alina M Allen, Zhen Wang, Larry J Prokop, Mohammad H Murad, Rohit Loomba
ABSTRACT
Little is known about differences in rates of fibrosis progression between patients with nonalcoholic fatty liver (NAFL) vs nonalcoholic steatohepatitis (NASH). We conducted a systematic review and meta-analysis of all studies that assessed paired liver biopsy specimens to estimate the rates of fibrosis progression in patients with nonalcoholic fatty liver disease (NAFLD) including NAFL and NASH. Through a systematic search of multiple databases and author contact, up to June 2013, we identified studies of adults with NAFLD that collected paired liver biopsy specimens at least 1 year apart. From these, we calculated a pooled-weighted annual fibrosis progression rate (number of stages changed between the 2 biopsy samples) with 95% confidence intervals (CIs), and identified clinical risk factors associated with progression. We identified 11 cohort studies including 411 patients with biopsy-proven NAFLD (150 with NAFL and 261 with NASH). At baseline, the distribution of fibrosis for stages 0, 1, 2, 3, and 4 was 35.8%, 32.5%, 16.7%, 9.3%, and 5.7%, respectively. Over 2145.5 person-years of follow-up evaluation, 33.6% had fibrosis progression, 43.1% had stable fibrosis, and 22.3% had an improvement in fibrosis stage. The annual fibrosis progression rate in patients with NAFL who had stage 0 fibrosis at baseline was 0.07 stages (95% CI, 0.02-0.11 stages), compared with 0.14 stages in patients with NASH (95% CI, 0.07-0.21 stages). These findings correspond to 1 stage of progression over 14.3 years for patients with NAFL (95% CI, 9.1-50.0 y) and 7.1 years for patients with NASH (95% CI, 4.8-14.3 y). Based on a meta-analysis of studies of paired liver biopsy studies, liver fibrosis progresses in patients with NAFL and NASH.
Product Description
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g