Merck
  • Mice with ablated adult brain neurogenesis are not impaired in antidepressant response to chronic fluoxetine.
Mice with ablated adult brain neurogenesis are not impaired in antidepressant response to chronic fluoxetine.
Journal of psychiatric research (2014-06-17)
Paulina Jedynak, Tomasz Kos, Carmen Sandi, Leszek Kaczmarek, Robert K Filipkowski
ABSTRACT
The neurogenesis hypothesis of major depression has two main facets. One states that the illness results from decreased neurogenesis while the other claims that the very functioning of antidepressants depends on increased neurogenesis. In order to verify the latter, we have used cyclin D2 knockout mice (cD2 KO mice), known to have virtually no adult brain neurogenesis, and we demonstrate that these mice successfully respond to chronic fluoxetine. After unpredictable chronic mild stress, mutant mice showed depression-like behavior in forced swim test, which was eliminated with chronic fluoxetine treatment, despite its lack of impact on adult hippocampal neurogenesis in cD2 KO mice. Our results suggest that new neurons are not indispensable for the action of antidepressants such as fluoxetine. Using forced swim test and tail suspension test, we also did not observe depression-like behavior in control cD2 KO mice, which argues against the link between decreased adult brain neurogenesis and major depression.
Product Description
Supelco
Fluoxetine hydrochloride, VETRANAL®, analytical standard
USP
Fluoxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluoxetine hydrochloride, solid
Supelco
Fluoxetine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material