Merck
  • NFAT2 regulates COX-2 expression and modulates the integrin repertoire in endothelial cells at the crossroads of angiogenesis and inflammation.
NFAT2 regulates COX-2 expression and modulates the integrin repertoire in endothelial cells at the crossroads of angiogenesis and inflammation.
Experimental cell research (2014-03-25)
Mari-Pau Mena, Izabela Papiewska-Pajak, Patrycja Przygodzka, Anna Kozaczuk, Joanna Boncela, Czeslaw S Cierniewski
ABSTRACT
The mechanisms controlling the switch between the pro-angiogenic and pro-inflammatory states of endothelial cells are still poorly understood. In this paper, we show that: (a) COX-2 expression induced by VEGF-A is NFAT2-dependent; and (b) the integrin profile in endothelial cells induced by the pro-angiogenic VEGF-A is distinct from that brought on by the inflammatory cytokine TNF-α. Two groups of integrin subunits specifically upregulated over time by both cytokines were identified using RT-PCR and Western Immunoblotting. The first group included α4, α5, α6, and β5 subunits that were upregulated by VEGF-A; the second group consisted of αV and β3 induced by TNF-α. Both cytokines significantly enhanced the expression of β1 and modulated α2 mRNA. In contrast to TNF-α, VEGF-A induction of integrin subunits depended on the activation of the calcineurin/NFAT pathway. Both calcineurin inhibitors (cyclosporineA and 11R-VIVIT) and downregulation of NFAT2 with specific siRNA decreased induction of integrin subunits. This process of induction could be increased by upregulation of NFAT2 by pBJ5-NFAT2 transfection. This suggests that NFAT2 mediates VEGF-induced upregulation of integrin subunit synthesis by providing a constant supply of newly synthesized "refreshed" mature integrin receptors, particularly α2β1, α5β1, α4β1, α6β1 and αVβ5, which are involved at different stages of angiogenesis.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Valdecoxib, ≥98% (HPLC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-CPP, solid