Merck
  • Determination of the maleic acid in rat urine and serum samples by isotope dilution-liquid chromatography-tandem mass spectrometry with on-line solid phase extraction.
Determination of the maleic acid in rat urine and serum samples by isotope dilution-liquid chromatography-tandem mass spectrometry with on-line solid phase extraction.
Talanta (2015-02-24)
Hsin-Chang Chen, Charlene Wu, Kuen-Yuh Wu
ABSTRACT
A rapid and simple on-line solid-phase extraction coupled with isotope dilution-liquid chromatography-tandem mass spectrometry (SPE-ID-LC-MS/MS) method was developed to quantitate maleic acid in serum and urine of SpragueDawley (SD) rats. The aforementioned biological samples were spiked with (13)C2-maleic acid, vigorously vortexed, added with acetonitrile to precipitate proteins, and then injected into the on-line SPE-LC-MS/MS system for quantification. Upon validation, this method demonstrated excellent feasibility and sensitivity: calibration curves for maleic acid in serum and urine display excellent linearity with the coefficient of determination (R(2)) greater than 0.999; the limits of detection and quantitation (LOD and LOQ) for maleic acid were determined at 0.2 and 0.5μg L(-1), respectively. Additionally, intra-day accuracy for maleic acid in serum and urine samples ranged from 94.0% to 100.2% and 101.3% to 104.4%, respectively. Furthermore, inter-day accuracy ranged from 93.6% to 101.0% and from 102.3% to 111.4% in serum and urine samples, respectively. Intra-day precision %RSD of maleic acid in serum and urine samples was 13.8% or less, whereas the inter-day precision was 6.1% or less. The matrix effects were not found to be statistically significant (p=0.9145 and p=0.5378, correspondingly) based on the calculations of recovery functions. The collected serum and urine samples were analyzed using SPE-ID-LC-MS/MS. Our results reveal trace levels of maleic acid in the control rats, demonstrating that this method is capable of analyzing background levels of contaminants in biofluids with excellent sensitivity and specificity at part-per-billion levels concentrations in complex matrices.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Maleic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Maleic acid, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)