Merck
  • Proinflammatory cytokines induce amelotin transcription in human gingival fibroblasts.
Proinflammatory cytokines induce amelotin transcription in human gingival fibroblasts.
Journal of oral science (2014-12-17)
Yohei Nakayama, Hideki Takai, Sari Matsui, Hiroyoshi Matsumura, Liming Zhou, Ayako Kato, Bernhard Ganss, Yorimasa Ogata
ABSTRACT
Amelotin (AMTN) is a secreted protein transcribed predominantly during the maturation stage of enamel formation and localized in the junctional epithelium. We investigated differences in the levels of AMTN gene expression between non-inflamed gingiva and inflamed gingiva from patients with chronic periodontitis. Total RNAs were isolated from these tissues and their gene expression profiles were monitored by DNA microarray. The observed induction of AMTN mRNA in inflamed gingiva and cultured human gingival fibroblasts (HGF) was confirmed by real-time PCR. Transient transfection assays were performed using chimeric constructs of mouse AMTN gene promoter fragments linked to a luciferase reporter gene. Immunohistochemical localization of AMTN in inflamed and non-inflamed gingiva was assessed by immunohistochemistry. Among many differentially expressed genes, the level of AMTN mRNA was significantly increased in inflamed gingiva. Treatment of HGF with interleukin-1β (IL-1β), IL-6 and tumor necrosis factor-α (TNF-α) induced the expression of AMTN mRNA, and increased the luciferase activities of the AMTN promoter constructs. AMTN protein was detected in inflamed gingival connective tissue and junctional epithelium. These findings demonstrate that proinflammatory cytokines induce AMTN gene expression in human gingival fibroblasts and suggest a role for AMTN in gingival inflammation.
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, for microscopy (Bact., Bot., Hist.)
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%