Merck
  • What the devil is in your phytomedicine? Exploring species substitution in Harpagophytum through chemometric modeling of 1H-NMR and UHPLC-MS datasets.
What the devil is in your phytomedicine? Exploring species substitution in Harpagophytum through chemometric modeling of 1H-NMR and UHPLC-MS datasets.
Phytochemistry (2014-07-22)
Nontobeko P Mncwangi, Alvaro M Viljoen, Jianping Zhao, Ilze Vermaak, Wei Chen, Ikhlas Khan
ABSTRACT
Harpagophytum procumbens (Pedaliaceae) and its close taxonomical ally Harpagophytum zeyheri, indigenous to southern Africa, are being harvested for exportation to Europe where phytomedicines are developed to treat inflammation-related disorders. The phytochemical variation within and between natural populations of H. procumbens (n=241) and H. zeyheri (n=107) was explored using proton nuclear magnetic resonance ((1)H-NMR) and ultra-high performance liquid chromatography coupled to mass spectrometry (UHPLC-MS) in combination with multivariate data analysis methods. The UHPLC-MS results revealed significant variation in the harpagoside content: H. procumbens (0.17-4.37%); H. zeyheri (0.00-3.07%). Only 41% of the H. procumbens samples and 17% of the H. zeyheri samples met the pharmacopoeial specification of ⩾1.2%. Both principal component analysis (PCA) and orthogonal projections to latent structures discriminant analysis (OPLS-DA) indicated separation based on species (UHPLC-MS data OPLS-DA model statistics: R(2)X=0.258, R(2)Y (cum)=0.957 and Q(2)(cum)=0.934; (1)H-NMR data OPLS-DA model statistics: R(2)X=0.830, R(2)Y=0.865 (cum) and Q(2)(cum)=0.829). It was concluded that two species are not chemically equivalent and should not be used interchangeably.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Harpagoside, primary reference standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Harpagoside, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Methanol, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis