Merck
  • Activatable hyaluronic acid nanoparticle as a theranostic agent for optical/photoacoustic image-guided photothermal therapy.
Activatable hyaluronic acid nanoparticle as a theranostic agent for optical/photoacoustic image-guided photothermal therapy.
ACS nano (2014-11-18)
Liwen Zhang, Shi Gao, Fan Zhang, Kai Yang, Qingjie Ma, Lei Zhu
ABSTRACT
Photothermal therapy (PTT) is an emerging treatment modality that is under intensive preclinical investigations for the treatment of various medical conditions, including cancer. However, the lack of targeting function of PTT agents hampers its clinical application. An effective and nontoxic delivery vehicle that can carry PTT agents into tumor areas is still needed urgently. In this study, we developed a multifunctional nanocomposite by loading copper sulfide (CuS) into Cy5.5-conjugated hyaluronic acid nanoparticles (HANP), obtaining an activatable Cy5.5-HANP/CuS (HANPC) nanocomposite. In this system, Cy5.5 fluorescent signal is quenched by CuS inside the particle until the whole nanocomposite is degraded by hyaluronidase present in tumor, giving strong fluorescence signals delineating the tumor. Importantly, CuS with strong NIR absorbance appears to be an excellent contrast agent for photoacoustic (PA) imaging and an effective PTT agent. After intravenous administration of HANPC into SCC7 tumor-bearing mice, high fluorescence and PA signals were observed in the tumor area over time, which peaked at the 6 h time point (tumor-to-normal tissue ratio of 3.25±0.25 for optical imaging and 3.8±0.42 for PA imaging). The tumors were then irradiated with a laser, and a good tumor inhibition rate (89.74% on day 5) was observed. Our studies further encourage application of this HA-based multifunctional nanocomposite for image-guided PTT in biomedical applications, especially in cancer theranostics.
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution