Merck
  • Steroid injection and needle aponeurotomy for Dupuytren disease: long-term follow-up of a randomized controlled trial.
Steroid injection and needle aponeurotomy for Dupuytren disease: long-term follow-up of a randomized controlled trial.
The Journal of hand surgery (2014-08-26)
Catherine McMillan, Paul Binhammer
ABSTRACT
To compare long-term outcomes and retreatment rates for patients with Dupuytren disease who underwent needle aponeurotomy (NA) combined with a series of triamcinolone acetonide injections or underwent NA alone as part of a prior randomized controlled trial. During this follow-up study, 44 of 47 participants in the original study were examined as needed between 6 and 53 months from their initial procedure. Those who had not been reassessed within 18 months of the original NA were asked to return for follow-up. The average total active extension deficit (TAED) of previously treated joints was compared between groups 7 to 12, 13 to 24, 25 to 36, and 37 to 48 months following treatment. Timing of retreatment (if performed) was recorded. Forty-four participants returned for assessment an average of 4.8 times over 53 months. Mean TAED was significantly less in needle aponeurotomy triamcinolone injection patients at 6 months and between 13 and 24 months. Sixty-two percent of NA group patients and 30% of needle aponeurotomy triamcinolone injection patients returned for a second treatment on the same digit(s) (retreatment). This difference was not significant. Mean time to retreatment and mean TAED immediately prior to retreatment did not differ significantly between groups. Kaplan-Meier survival estimates demonstrated a significantly higher percentage of NA group patients expected to return for retreatment by 24 but not by 36 months. Younger age, more than one joint treated at the initial NA, and TAED severity throughout the follow-up period were associated with earlier retreatment. Serial triamcinolone injections combined with NA was associated with lower TAED for up to 24 months. A larger study would more accurately quantify the potential benefits of combining triamcinolone injections with NA for treatment of Dupuytren disease. Therapeutic III.
Product Description
Supelco
Triamcinolone acetonide, analytical standard
Triamcinolone acetonide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Triamcinolone acetonide, United States Pharmacopeia (USP) Reference Standard
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard